Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyltri-n-butylammonium iodide, 97%

$65.57 - $204.73

Chemical Identifiers

Chemical Name or Material Benzyltri-n-butylammonium iodide
CAS 60754-76-9
Molecular Formula [CH3(CH2)3]3N(CH2C6H5)I
Formula Weight 403.39
MDL Number MFCD00011850
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2283714

 
alfa aesar™
B2283714
25g Each for $65.57

AAB2283722

 
alfa aesar™
B2283722
100g Each for $204.73
Description & Specifications

Specifications

Formula Weight 403.39
Chemical Name or Material Benzyltri-n-butylammonium iodide
Assay 97%
Molecular Formula [CH3(CH2)3]3N(CH2C6H5)I
MDL Number MFCD00011850
CAS 60754-76-9
Melting Point 143° to 145°C
Health Hazard 1 Danger
Quantity 25g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.