Learn More
Benzyltri-n-butylammonium chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A103450E
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzyltri-n-butylammonium chloride | |
156°C to 162°C | |
MFCD00011849 | |
3776210 | |
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac | |
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1 | |
311.94 | |
311.94 |
23616-79-7 | |
C19H34ClN | |
2500 g | |
Hygroscopic | |
VJGNLOIQCWLBJR-UHFFFAOYSA-M | |
benzyl(tributyl)azanium;chloride | |
159952 | |
98% |
Chemical Identifiers
23616-79-7 | |
311.94 | |
VJGNLOIQCWLBJR-UHFFFAOYSA-M | |
159952 | |
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1 |
C19H34ClN | |
MFCD00011849 | |
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac | |
benzyl(tributyl)azanium;chloride |
Safety and Handling
GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 245-787-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only