Please login to your online account to display your discounted pricing

Alfa Aesar™ (Benzylthio)acetic acid, 97%

$56.50 - $197.39

Chemical Identifiers

Chemical Name or Material (Benzylthio)acetic acid
CAS 103-46-8
Molecular Formula C6H5CH2SCH2CO2H
Formula Weight 182.24
MDL Number MFCD00004358
Beilstein 2047546
Synonym S-Benzylthioglycolic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1869809

  SDS
alfa aesar™
A1869809
10g Each for $56.50

AAA1869818

  SDS
alfa aesar™
A1869818
50g Each for $197.39
Description & Specifications

Specifications

Formula Weight 182.24
TSCA TSCA
Assay 97%
Odor Stench
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 204°C (27mmHg)
CAS 103-46-8
Health Hazard 1 Warning
Chemical Name or Material (Benzylthio)acetic acid
Melting Point 60° to 63°C
Molecular Formula C6H5CH2SCH2CO2H
Beilstein 2047546
Synonym S-Benzylthioglycolic acid
Quantity 10g
MDL Number MFCD00004358
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.