missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyloxyacetic Acid, TCI America™

$48.89 - $122.22

Chemical Identifiers

CAS 30379-55-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00274211
InChI Key GRZHHTYDZVRPIC-UHFFFAOYSA-N
Synonym benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy, benzyloxy acetic acid
PubChem CID 290301
IUPAC Name 2-phenylmethoxyacetic acid
SMILES C1=CC=C(C=C1)COCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28761G View Documents TCI America
B28761G
1g Each for $48.89
 
B28765G View Documents TCI America
B28765G
5g Each for $122.22
 
Specifications

Chemical Identifiers

30379-55-6
166.176
GRZHHTYDZVRPIC-UHFFFAOYSA-N
290301
C1=CC=C(C=C1)COCC(=O)O
C9H10O3
MFCD00274211
benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy, benzyloxy acetic acid
2-phenylmethoxyacetic acid

Specifications

155°C
1g
C9H10O3
benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy, benzyloxy acetic acid
C1=CC=C(C=C1)COCC(=O)O
166.176
166.18
≥97.0% (GC,T)
Yellow
30379-55-6
MFCD00274211
GRZHHTYDZVRPIC-UHFFFAOYSA-N
2-phenylmethoxyacetic acid
290301
Clear Liquid at 20°C
Benzyloxyacetic Acid
Safety and Handling

Safety and Handling

TSCA : No