missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyloxyacetic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 427080050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Benzyloxyacetic Acid | |
1.1620g/mL | |
113°C | |
Glass Bottle | |
C6H5CH2OCH2COOH | |
5g | |
benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy | |
OC(=O)COCC1=CC=CC=C1 | |
166.18 | |
166.18 |
30379-55-6 | |
137.0°C to 139.0°C (0.6mmHg) | |
94% min. (HPLC) | |
C9H10O3 | |
MFCD00274211 | |
1.162 | |
GRZHHTYDZVRPIC-UHFFFAOYSA-N | |
2-phenylmethoxyacetic acid | |
290301 | |
95% |
Chemical Identifiers
30379-55-6 | |
166.18 | |
GRZHHTYDZVRPIC-UHFFFAOYSA-N | |
290301 | |
OC(=O)COCC1=CC=CC=C1 |
C9H10O3 | |
MFCD00274211 | |
benzyloxyacetic acid, 2-benzyloxy acetic acid, benzyloxy acetic acid, acetic acid, phenylmethoxy, phenylmethoxyaceticacid, 2-phenylmethoxy acetic acid, benzyloxy-acetic acid, 2-benzyloxyacetic acid, acetic acid, 2-phenylmethoxy | |
2-phenylmethoxyacetic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only