Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyloxyacetaldehyde dimethyl acetal, 97%

$103.35 - $378.20

Chemical Identifiers

Chemical Name or Material Benzyloxyacetaldehyde dimethyl acetal
CAS 127657-97-0
Molecular Formula C6H5CH2OCH2CH(OCH3)2
Formula Weight 196.24
MDL Number MFCD00082233
Beilstein 3261948
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0865109

  SDS
alfa aesar™
L0865109
10g Each for $103.35

AAL0865118

  SDS
alfa aesar™
L0865118
50g Each for $378.20
Description & Specifications

Specifications

Formula Weight 196.24
Assay 97%
Chemical Name or Material Benzyloxyacetaldehyde dimethyl acetal
CAS 127657-97-0
Molecular Formula C6H5CH2OCH2CH(OCH3)2
MDL Number MFCD00082233
Beilstein 3261948
Quantity 10g
Boiling Point 80° to 82°C (0.5mmHg)