Learn More
Benzylmagnesium chloride, 1-2M in THF, Thermo Scientific Chemicals
$149.48 - $149.48
Chemical Identifiers
CAS | 6921-34-2 |
---|---|
Molecular Formula | C7H7ClMg |
Molecular Weight (g/mol) | 150.89 |
MDL Number | MFCD00000469 |
InChI Key | NRAFPLGJPPJUNB-UHFFFAOYSA-M |
Synonym | benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium |
PubChem CID | 2733352 |
IUPAC Name | magnesium;methanidylbenzene;chloride |
SMILES | Cl[Mg]CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA87299AD
|
Thermo Scientific Chemicals
087299AD |
50 mL |
Each for $149.48
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6921-34-2 | |
150.89 | |
NRAFPLGJPPJUNB-UHFFFAOYSA-M | |
2733352 | |
Cl[Mg]CC1=CC=CC=C1 |
C7H7ClMg | |
MFCD00000469 | |
benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium | |
magnesium;methanidylbenzene;chloride |
Specifications
6921-34-2 | |
C7H7ClMg | |
50 mL | |
Air and moisture sensitive | |
NRAFPLGJPPJUNB-UHFFFAOYSA-M | |
magnesium;methanidylbenzene;chloride | |
2733352 | |
1 to 2M in THF | |
Benzylmagnesium chloride |
Brown to Purple | |
MFCD00000469 | |
UN2924 | |
benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium | |
Cl[Mg]CC1=CC=CC=C1 | |
150.89 | |
150.89 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335-H336-H351
DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only