Please login to your online account to display your discounted pricing

Benzylidenemalononitrile, 98%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  164290250

 View more versions of this product

Catalog No. AC164290250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Needle-Like Crystalline Powder
Assay ≥97.5%
Formula Weight 154.17
Quantity 25g
Color Slightly yellow
Beilstein 09, 895
EINECS Number 220-283-6
RTECS Number OO3500000
Melting Point 82° to 86°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C10H6N2
Chemical Name or Material Benzylidenemalononitrile
Assay Percent Range 98%
MDL Number MFCD00001855
CAS Min % 97.5
Linear Formula C6H5CH=C(CN)2
Name Note 98%
CAS 2700-22-3
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Benzylidenemalononitrile
Name Note 98%
CAS 2700-22-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H6N2
Linear Formula C6H5CH=C(CN)2
Formula Weight 154.17
MDL Number MFCD00001855
Accessories

GHS Signal Word: Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents