missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzylidenemalononitrile, 98%, ACROS Organics™

Chemical Identifiers

CAS 2700-22-3
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.172
MDL Number MFCD00001855
InChI Key WAVNYPVYNSIHNC-UHFFFAOYSA-N
Synonym 2,2-dicyano-1-phenylethylene, 2-benzylidenemalononitrile, 2-phenyl-1,1-dicyanoethylene, benzal-malonitril, benzalmalononitrile, benzylidene malononitrile, benzylidenemalonodinitrile, benzylidenemalononitrile, beta,beta-dicyanostyrene, malononitrile, benzylidene
PubChem CID 17608
IUPAC Name 2-benzylidenepropanedinitrile
SMILES C1=CC=C(C=C1)C=C(C#N)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC164291000
SDS View Documents
Thermo Fisher Scientific
164291000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2700-22-3
154.172
WAVNYPVYNSIHNC-UHFFFAOYSA-N
17608
C1=CC=C(C=C1)C=C(C#N)C#N
C10H6N2
MFCD00001855
2,2-dicyano-1-phenylethylene, 2-benzylidenemalononitrile, 2-phenyl-1,1-dicyanoethylene, benzal-malonitril, benzalmalononitrile, benzylidene malononitrile, benzylidenemalonodinitrile, benzylidenemalononitrile, beta,beta-dicyanostyrene, malononitrile, benzylidene
2-benzylidenepropanedinitrile

Specifications

Authentic
Yellow
100g
97.5
98%
C6H5CH=C(CN)2
09, 895
WAVNYPVYNSIHNC-UHFFFAOYSA-N
2-benzylidenepropanedinitrile
17608
Needle-Like Crystalline Powder
Benzylidenemalononitrile, 98%
Glass Bottle
82°C to 86°C
2700-22-3
100.0
C10H6N2
MFCD00001855
2,2-dicyano-1-phenylethylene, 2-benzylidenemalononitrile, 2-phenyl-1,1-dicyanoethylene, benzal-malonitril, benzalmalononitrile, benzylidene malononitrile, benzylidenemalonodinitrile, benzylidenemalononitrile, beta,beta-dicyanostyrene, malononitrile, benzylidene
C1=CC=C(C=C1)C=C(C#N)C#N
154.172
154.17
≥97.5%
Safety and Handling

Safety and Handling

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 220-283-6

RTECSNumber : OO3500000

SDS