missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzylhydrazine dihydrochloride, 97%

Manufacturer:  Alfa Aesar™ H5491614

 View more versions of this product

Catalog No. AAH5491614



Description

Description

Applications
Benzylhydrazine dihydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Benzylhydrazine dihydrochloride
25g
C7H12Cl2N2
MFCD00012921
3688990
Insoluble in water.
C1=CC=C(C=C1)CNN.Cl.Cl
195.087
195.09
143°C to 145°C
20570-96-1
C6H5CH2NHNH2·2HCl
UN2811
1-benzylhydrazine dihydrochloride
MSJHOJKVMMEMNX-UHFFFAOYSA-N
benzylhydrazine;dihydrochloride
146540
97%

Chemical Identifiers

20570-96-1
195.087
MSJHOJKVMMEMNX-UHFFFAOYSA-N
146540
C1=CC=C(C=C1)CNN.Cl.Cl
C7H12Cl2N2
MFCD00012921
1-benzylhydrazine dihydrochloride
benzylhydrazine;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 243-887-1

RTECSNumber : MU8540000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents