Please login to your online account to display your discounted pricing

Benzyldiphenylphosphine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzyldiphenylphosphine
CAS 7650-91-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C19H17P
Linear Formula C6H5CH2P(C6H5)2
Formula Weight 276.32
MDL Number MFCD00014083
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378840010

  SDS
thermo fisher scientific
AC378840010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 276.32
Quantity 1g
Color White to off-white
CAS Max % 100.0
CAS Min % 96.0
Molecular Formula C19H17P
Infrared Spectrum Authentic
Chemical Name or Material Benzyldiphenylphosphine
Assay Percent Range 97%
MDL Number MFCD00014083
Packaging Glass bottle
Linear Formula C6H5CH2P(C6H5)2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 7650-91-1
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.