missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyldimethylhexadecylammonium chloride hydrate, 95%

Manufacturer:  Alfa Aesar™ 3237718

 View more versions of this product

Catalog No. AA3237718



Specifications

Specifications

Benzyldimethylhexadecylammonium chloride hydrate
54°C to 58°C
122-18-9
CH3(CH2)15N(CH2C6H5)(CH3)2Cl·xH2O
UN3261
14,2017
SXPWTBGAZSPLHA-UHFFFAOYSA-M
benzyl-hexadecyl-dimethylazanium;chloride
31202
Powder
White
50g
C25H46ClN
MFCD00011625
3802341
cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]
396.1
396.10 (Anhydrous)
95%

Chemical Identifiers

122-18-9
396.1
SXPWTBGAZSPLHA-UHFFFAOYSA-M
31202
CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]
C25H46ClN
MFCD00011625
cetalkonium chloride, benzyldimethylhexadecylammonium chloride, banicol, bonjela, cetyl zephiran, cetyldimethylbenzylammonium chloride, bicetonium, acinol, benzylhexadecyldimethylammonium chloride, baktonium
benzyl-hexadecyl-dimethylazanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : BO6822450

TSCA : TSCA

SDS Documents