Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzylamine on polystyrene, 2.0-3.0 mmol/g

$118.89 - $453.54

Chemical Identifiers

Chemical Name or Material Benzylamine on polystyrene
CAS 89551-24-6
MDL Number MFCD00130502
Synonym Aminomethylated polystyrene; AM resin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1947106

  SDS
alfa aesar™
L1947106
5g Each for $118.89

AAL1947114

  SDS
alfa aesar™
L1947114
25g Each for $453.54
Description & Specifications

Specifications

MDL Number MFCD00130502
Chemical Name or Material Benzylamine on polystyrene
CAS 89551-24-6
Synonym Aminomethylated polystyrene; AM resin
Concentration 2.0 to 3.0mmol/g
Quantity 5g