Please login to your online account to display your discounted pricing

Benzylamine hydrochloride, 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  200541000

 View more versions of this product

Catalog No. AC200541000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 143.62
Quantity 100g
Color White
Beilstein 12, 1017
Merck Index 14, 1125
EINECS Number 221-943-6
TSCA TSCA
Melting Point 259° to 264°C
RTECS Number DP5425000
CAS Max % 100.0
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material Benzylamine hydrochloride
Assay Percent Range 99%
Molecular Formula C7H9N·ClH
MDL Number MFCD00012852
Name Note 99%
Linear Formula C6H5CH2NH2·HCl
CAS 3287-99-8
Infrared Spectrum Authentic
Synonym Benzylammonium chloride
CAS Min % 98.5
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Benzylamine hydrochloride
Name Note 99%
CAS 3287-99-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H9N·ClH
Linear Formula C6H5CH2NH2·HCl
Formula Weight 143.62
MDL Number MFCD00012852
Synonym Benzylammonium chloride
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents