Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzylamine hydrochloride, 99%

$209.45 - $209.45

Chemical Identifiers

Chemical Name or Material Benzylamine hydrochloride
CAS 3287-99-8
Molecular Formula C7H9N·HCl
Formula Weight 143.62
MDL Number MFCD00012852
Beilstein 3594394
Merck Index 14,1125
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0410236

  SDS
alfa aesar™
L0410236
500g Each for $209.45
Description & Specifications

Specifications

Formula Weight 143.62
MDL Number MFCD00012852
Chemical Name or Material Benzylamine hydrochloride
Assay 99%
CAS 3287-99-8
Health Hazard 1 Danger
Molecular Formula C7H9N·HCl
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,1125
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 262° to 263°C
Beilstein 3594394
TSCA TSCA
RTECS Number DP5425000
Quantity 500g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.