missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzylamine, +99.5%, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427231000

 View more versions of this product

Catalog No. AC427231000


 

Specifications Description & Specifications

Specifications

Benzylamine
99.5
Liquid
99.5+%
C6H5CH2NH2
MFCD00008106
15, 1128
AcroSeal™ Glass Bottle
0.3% max.
Clear colorless to slightly yellow
100-46-9
100.0
≥99.50%
C7H9N
107.16
Authentic
Benzenemethanamine
1.5424 to 1.5444 (20°C, 589nm)
0.9800g/mL
100mL

Chemical Identifiers

100-46-9
100.0
C7H9N
107.16
Benzenemethanamine
≥99.50%
99.5
99.5+%
C6H5CH2NH2
MFCD00008106
Benzylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger