Please login to your online account to display your discounted pricing

Benzylacetone 98%, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC105830010

 View more versions of this product

Catalog No. AC105830010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 148.2
Physical Form Liquid
Quantity 1L
Color Clear colorless to pale yellow
EINECS Number 219-847-4
TSCA TSCA
RTECS Number EL9600000
Density 0.9875g/mL
Specific Gravity 0.985 to 0.99 (25°/25°C)
Beilstein 07, 314
CAS Max % 100.0
Synonym 4-Phenyl-2-butanone
CAS 2550-26-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
CAS Min % 98.5
Linear Formula C6H5CH2CH2COCH3
Chemical Name or Material Benzylacetone
Molecular Formula C10H12O
MDL Number MFCD00008790
Name Note 98%
Refractive Index 1.5100 to 1.5130 (20°C, 589nm)
Packaging Glass bottle

Specifications

Chemical Name or Material Benzylacetone
Name Note 98%
CAS 2550-26-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H12O
Linear Formula C6H5CH2CH2COCH3
Formula Weight 148.2
MDL Number MFCD00008790
Synonym 4-Phenyl-2-butanone
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.

SDS & Safety Documents