missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzylacetone, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 105832500

 View more versions of this product

Catalog No. AC105832500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Benzylacetone
105°C
Glass bottle
0.9875
Colorless to Yellow
98%
98.5
98.5% min. (GC)
C6H5CH2CH2COCH3
07, 314
Solubility in water: practically insoluble in water.. Other solubilities: soluble in ethanol,diethyl ether,chloroform,,very soluble in acetone
CC(=O)CCC1=CC=CC=C1
148.2
148.2
99%
0.9875g/mL
Authentic
1.5100 to 1.513
235°C to 237°C
250mL
2550-26-7
100.0
C10H12O
MFCD00008790
1-phenyl-3-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 4-phenyl-2-butanone, benzyl acetone, benzylacetone, methyl 2-phenylethyl ketone, methyl phenethyl ketone, methyl phenylethyl ketone, phenethyl methyl ketone
AKGGYBADQZYZPD-UHFFFAOYSA-N
4-phenylbutan-2-one
17355
Liquid

Chemical Identifiers

2550-26-7
148.2
AKGGYBADQZYZPD-UHFFFAOYSA-N
17355
CC(=O)CCC1=CC=CC=C1
C10H12O
MFCD00008790
1-phenyl-3-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 4-phenyl-2-butanone, benzyl acetone, benzylacetone, methyl 2-phenylethyl ketone, methyl phenethyl ketone, methyl phenylethyl ketone, phenethyl methyl ketone
4-phenylbutan-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-847-4

RTECSNumber : EL9600000

TSCA : TSCA

SDS Documents