Please login to your online account to display your discounted pricing

Benzylacetone 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  105830050

 View more versions of this product

Catalog No. AC105830050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 148.2
Quantity 5mL
Color Clear colorless to pale yellow
Beilstein 07, 314
TSCA TSCA
RTECS Number EL9600000
EINECS Number 219-847-4
Specific Gravity 0.985 to 0.99 (25°/25°C)
CAS Max % 100.0
Refractive Index 1.5100 to 1.5130 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Linear Formula C6H5CH2CH2COCH3
MDL Number MFCD00008790
CAS 2550-26-7
Assay Percent Range 99%
Name Note 98%
CAS Min % 98.5
Molecular Formula C10H12O
Chemical Name or Material Benzylacetone
Synonym 4-Phenyl-2-butanone
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Benzylacetone
Name Note 98%
CAS 2550-26-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H12O
Linear Formula C6H5CH2CH2COCH3
Formula Weight 148.2
MDL Number MFCD00008790
Synonym 4-Phenyl-2-butanone
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.

SDS & Safety Documents