Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl thiocyanate, 98+%

Chemical Identifiers

Chemical Name or Material Benzyl thiocyanate
CAS 3012-37-1
Molecular Formula C6H5CH2SCN
Formula Weight 149.22
MDL Number MFCD00001832
UN Number UN2811
Beilstein 1859726
Synonym Benzyl rhodanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2390230

  SDS
alfa aesar™
B2390230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2390218

  SDS
alfa aesar™
B2390218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 149.22
Chemical Name or Material Benzyl thiocyanate
TSCA TSCA
Assay ≥98%
Molecular Formula C6H5CH2SCN
Melting Point 40° to 43°C
MDL Number MFCD00001832
CAS 3012-37-1
UN Number UN2811
Beilstein 1859726
Boiling Point 230°C
Quantity 250g
Flash Point 110°C (230°F)
Synonym Benzyl rhodanide
Odor Irritating
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number XK8155000
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.