missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5184414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzyl thioacetimidate hydrochloride | |
C9H12ClNS | |
25 g | |
OVQWHRFBVPDPCD-UHFFFAOYSA-N | |
benzyl ethanimidothioate;hydrochloride | |
12626231 | |
96% |
32894-07-8 | |
MFCD05865210 | |
s-benzylthioacetimidate, hydrochloride, benzyl thioacetimidate hydrochloride, benzyl thioacetimidate hcl, benzyl ethanimidothioate hydrochloride, 1-benzylsulfanyl ethanimine hydrochloride, acmc-20ajr8, s-benzylthioacetimidate hydrochloride, thioacetimidic acid benzyl ester; hydrochloride, benzyl ethanimidothioate-hydrogen chloride 1/1 | |
CC(=N)SCC1=CC=CC=C1.Cl | |
201.712 | |
201.71 |
Chemical Identifiers
32894-07-8 | |
201.712 | |
OVQWHRFBVPDPCD-UHFFFAOYSA-N | |
12626231 | |
CC(=N)SCC1=CC=CC=C1.Cl |
C9H12ClNS | |
MFCD05865210 | |
s-benzylthioacetimidate, hydrochloride, benzyl thioacetimidate hydrochloride, benzyl thioacetimidate hcl, benzyl ethanimidothioate hydrochloride, 1-benzylsulfanyl ethanimine hydrochloride, acmc-20ajr8, s-benzylthioacetimidate hydrochloride, thioacetimidic acid benzyl ester; hydrochloride, benzyl ethanimidothioate-hydrogen chloride 1/1 | |
benzyl ethanimidothioate;hydrochloride |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only