Please login to your online account to display your discounted pricing

Benzyl sulfoxide 98+%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC106175000

 View more versions of this product

Catalog No. AC106175000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 230.32
Physical Form Crystalline Powder
Melting Point >130°C
Quantity 500g
Color Off-white to pale yellow
Beilstein 06, 456
CAS Min % 98.0
CAS Max % 100.0
Molecular Formula C14H14OS
Linear Formula (C6H5CH2)2SO
MDL Number MFCD00004782
CAS 621-08-9
Synonym Dibenzyl sulfoxide
Chemical Name or Material Benzyl sulfoxide
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98+%

Specifications

Chemical Name or Material Benzyl sulfoxide
CAS 621-08-9
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C14H14OS
Linear Formula (C6H5CH2)2SO
Formula Weight 230.32
MDL Number MFCD00004782
Synonym Dibenzyl sulfoxide