missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl sulfoxide 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC106171000

 View more versions of this product

Catalog No. AC106171000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.0%
230.32
100g
06, 456
100.0
Authentic
MFCD00004782
(C6H5CH2)2SO
98+%
>130°C
Crystalline Powder
Off-white to pale yellow
98.0
Glass bottle
Dibenzyl sulfoxide
621-08-9
C14H14OS
Benzyl sulfoxide

Chemical Identifiers

621-08-9
100.0
C14H14OS
230.32
Dibenzyl sulfoxide
≥98.0%
98.0
98+%
(C6H5CH2)2SO
MFCD00004782
Benzyl sulfoxide