missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl sulfoxide 98+%, ACROS Organics™

Chemical Identifiers

CAS 621-08-9
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C14H14OS
Linear Formula (C6H5CH2)2SO
Formula Weight 230.32
MDL Number MFCD00004782
Synonym Dibenzyl sulfoxide
Chemical Name or Material Benzyl sulfoxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC106175000 SDS Thermo Fisher Scientific
AC106175000
Glass bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC106170250 SDS Thermo Fisher Scientific
AC106170250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC106171000 SDS Thermo Fisher Scientific
AC106171000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

621-08-9
100.0
C14H14OS
230.32
Dibenzyl sulfoxide
≥98.0%
98.0
98+%
(C6H5CH2)2SO
MFCD00004782
Benzyl sulfoxide

Specifications

230.32
>130°C
Off-white to pale yellow
98.0
Dibenzyl sulfoxide
(C6H5CH2)2SO
MFCD00004782
Authentic
Benzyl sulfoxide
Crystalline Powder
500g
06, 456
100.0
621-08-9
C14H14OS
Glass bottle
98+%
≥98.0%