missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC434190050

 View more versions of this product

Catalog No. AC434190050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate
Glass bottle
White
5g
96.0
97%
MFCD00216569
BNIBNUOPVTZWRT-JTQLQIEISA-N
benzyl N-[(3S)-5-oxooxolan-3-yl]carbamate
697922
Crystalline Powder
Authentic
−47° to −57° (20°C, 589nm) (C=1, acetone)
105.0°C to 106.5°C
87219-29-2
100.0
C12H13NO4
benzyl s---tetrahydro-5-oxo-3-furanylcarbamate, s-benzyl 5-oxotetrahydrofuran-3-yl carbamate, benzyl n-3s-5-oxooxolan-3-yl carbamate, benzyl s---tetrahydro-5-oxo-3-furanyl-carbama, s-benzyl-5-oxo-tetrahydro-furan-3-ylcarbamate, benzyl 3s-5-oxotetrahydrofuran-3-ylcarbamate, benzyl s-tetrahydro-5-oxo-3-furanylcarbamate, n-3s-5-oxo 3-2,3,4-trihydrofuryl phenylmethoxy carboxamide, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester 9ci
C1C(COC1=O)NC(=O)OCC2=CC=CC=C2
235.239
235.24
97%

Chemical Identifiers

87219-29-2
235.239
BNIBNUOPVTZWRT-JTQLQIEISA-N
697922
C1C(COC1=O)NC(=O)OCC2=CC=CC=C2
C12H13NO4
MFCD00216569
benzyl s---tetrahydro-5-oxo-3-furanylcarbamate, s-benzyl 5-oxotetrahydrofuran-3-yl carbamate, benzyl n-3s-5-oxooxolan-3-yl carbamate, benzyl s---tetrahydro-5-oxo-3-furanyl-carbama, s-benzyl-5-oxo-tetrahydro-furan-3-ylcarbamate, benzyl 3s-5-oxotetrahydrofuran-3-ylcarbamate, benzyl s-tetrahydro-5-oxo-3-furanylcarbamate, n-3s-5-oxo 3-2,3,4-trihydrofuryl phenylmethoxy carboxamide, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester 9ci
benzyl N-[(3S)-5-oxooxolan-3-yl]carbamate
SDS Documents