Please login to your online account to display your discounted pricing

Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate
CAS 87219-29-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C12H13NO4
Formula Weight 235.24
MDL Number MFCD00216569
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC434190010

  SDS
thermo fisher scientific
AC434190010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434190050

  SDS
thermo fisher scientific
AC434190050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0% (HPLC)
Formula Weight 235.24
Quantity 1g
Color White
Melting Point 103° to 108°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Specific Rotation −47° to −57° (20°C, 589nm) (C=1, acetone)
Chemical Name or Material Benzyl (S)-(-)-tetrahydro-5-oxo-3-furanylcarbamate
Molecular Formula C12H13NO4
CAS 87219-29-2
Assay Percent Range 97%
MDL Number MFCD00216569