Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%

Chemical Identifiers

Chemical Name or Material Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
CAS 113400-36-5
Formula Weight 319.35
MDL Number MFCD04115781
Synonym Boc-Pyr-Obzl; N-Boc-L-pyroglutamic acid benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6291203

  SDS
alfa aesar™
H6291203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6291206

  SDS
alfa aesar™
H6291206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62912MD

  SDS
alfa aesar™
H62912MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 319.35
Physical Form Powder
Color White
Chemical Name or Material Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
Assay 98%
Quantity 1g
MDL Number MFCD04115781
Synonym Boc-Pyr-Obzl; N-Boc-L-pyroglutamic acid benzyl ester
CAS 113400-36-5