missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl (R)-(-)-glycidyl ether, 98+%

Manufacturer:  Alfa Aesar™ L1398003

 View more versions of this product

Catalog No. AAL1398003



Description

Description

Applications
Benzyl (R)-(-)-glycidyl ether is used in the preparation of the lactone fragment of compactin and mevinolin. Chiron in the preparation of syn-1,3-polyols, dideoxynucleosides, and a spiroacetal cyanohydrin.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Specifications

Benzyl (R)-(-)-glycidyl ether
−5.1° (c=5 in Toluene)
1g
C10H12O2
3588399
QNYBOILAKBSWFG-JTQLQIEISA-N
(2R)-2-(phenylmethoxymethyl)oxirane
159746
≥98%
>110°C (230°F)
130°C (0.1mmHg)
14618-80-5
MFCD00068409
r-benzyloxymethyl-oxirane, r---benzyl glycidyl ether, r-2-benzyloxy methyl oxirane, benzyl r---glycidyl ether, r-o-benzylglycidol, r-benzyl glycidyl ether, r---2-benzyloxymethyl oxirane, ccris 6385, --benzyl-r-glycidylether, r---benzyloxymethyloxirane
C1C(O1)COCC2=CC=CC=C2
164.204
164.2

Chemical Identifiers

14618-80-5
164.204
QNYBOILAKBSWFG-JTQLQIEISA-N
159746
C1C(O1)COCC2=CC=CC=C2
C10H12O2
MFCD00068409
r-benzyloxymethyl-oxirane, r---benzyl glycidyl ether, r-2-benzyloxy methyl oxirane, benzyl r---glycidyl ether, r-o-benzylglycidol, r-benzyl glycidyl ether, r---2-benzyloxymethyl oxirane, ccris 6385, --benzyl-r-glycidylether, r---benzyloxymethyloxirane
(2R)-2-(phenylmethoxymethyl)oxirane
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P281-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TX2860030

Documents