missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl propargyl ether, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5348114

 View more versions of this product

Catalog No. AAH5348114



Description

Description

Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl propargyl ether is used in the synthesis of substituted carbocyclic aromatic compounds and pharmaceutical compounds.

Solubility
Soluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Benzyl propargyl ether
1.011
C10H10O
1.52
25 g
Soluble in chloroform.
C#CCOCC1=CC=CC=C1
146.189
146.19
4039-82-1
101°C to 101°C (18 mmHg)
C6H4CH2OCH2C≡CH
MFCD00483987
benzyl propargyl ether, benzylpropargyl ether, 1-benzyloxy-2-propyne, prop-2-yn-1-yloxy methyl benzene, benzene, 2-propynyloxy methyl, prop-2-ynyloxy methyl benzene, propargylbenzyl ether, benzyloxy prop-2-yne
PAQVEXAFKDWGOT-UHFFFAOYSA-N
prop-2-ynoxymethylbenzene
6917484
97%

Chemical Identifiers

4039-82-1
146.189
PAQVEXAFKDWGOT-UHFFFAOYSA-N
6917484
C#CCOCC1=CC=CC=C1
C10H10O
MFCD00483987
benzyl propargyl ether, benzylpropargyl ether, 1-benzyloxy-2-propyne, prop-2-yn-1-yloxy methyl benzene, benzene, 2-propynyloxy methyl, prop-2-ynyloxy methyl benzene, propargylbenzyl ether, benzyloxy prop-2-yne
prop-2-ynoxymethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only