Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl phenyl sulfone, 98+%

$91.73 - $354.04

Chemical Identifiers

Chemical Name or Material Benzyl phenyl sulfone
CAS 3112-88-7
Molecular Formula C13H12O2S
Formula Weight 232.29
MDL Number MFCD00025040
Beilstein 2049938
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0420109

  SDS
alfa aesar™
L0420109
10g Each for $91.73

AAL0420118

  SDS
alfa aesar™
L0420118
50g Each for $354.04
Description & Specifications

Specifications

Formula Weight 232.29
Chemical Name or Material Benzyl phenyl sulfone
Molecular Formula C13H12O2S
MDL Number MFCD00025040
CAS 3112-88-7
Beilstein 2049938
Quantity 10g
Assay ≥98%
Melting Point 147° to 150°C
Odor Odorless
RTECS Number WR2700000