missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl phenyl sulfide, 98%

$193.95 - $785.03

Chemical Identifiers

CAS 831-91-4
Molecular Formula C6H5CH2SC6H5
Formula Weight 200.3
MDL Number MFCD00003066
Beilstein 1909409
Chemical Name or Material Benzyl phenyl sulfide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1617122 SDS Alfa Aesar™
A1617122
100g Each for $193.95
AAA1617136 SDS Alfa Aesar™
A1617136
500g Each for $785.03
AAA1617114 SDS Alfa Aesar™
A1617114
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Benzyl phenyl sulfide is used in dyestuff intermediate, flavor & fragrance intermediate and organic intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

831-91-4
200.3
1909409
C6H5CH2SC6H5
MFCD00003066
Benzyl phenyl sulfide

Specifications

831-91-4
200.3
MFCD00003066
167° to 168°C (10mmHg)
>110°C (230°F)
Benzyl phenyl sulfide
C6H5CH2SC6H5
39° to 42°C
1909409
100g
Stench
98%