Please login to your online account to display your discounted pricing

Benzyl phenyl ether, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC310260050

 View more versions of this product

Catalog No. AC310260050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Chunks
Assay ≥97.5%
Formula Weight 184.24
Quantity 5g
Color Light brown-orange
Melting Point 37° to 41°C
CAS Max % 100.0
Linear Formula C6H5CHOC6H5
Assay Percent Range 98%
Molecular Formula C13H12O
CAS 946-80-5
Chemical Name or Material Benzyl phenyl ether
Infrared Spectrum Authentic
MDL Number MFCD00020660
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Benzyl phenyl ether
CAS 946-80-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H12O
Linear Formula C6H5CHOC6H5
Formula Weight 184.24
MDL Number MFCD00020660
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents