missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl Nicotinate, TCI America™

$64.17 - $149.72

Chemical Identifiers

CAS 94-44-0
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00023584
InChI Key KVYGGMBOZFWZBQ-UHFFFAOYSA-N
Synonym benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
PubChem CID 7191
ChEBI CHEBI:31268
IUPAC Name benzyl pyridine-3-carboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N008325G View Documents TCI America
N008325G
25g Each for $64.17
 
N0083100G View Documents TCI America
N0083100G
100g Each for $149.72
 
Specifications

Chemical Identifiers

94-44-0
213.236
KVYGGMBOZFWZBQ-UHFFFAOYSA-N
7191
benzyl pyridine-3-carboxylate
C13H11NO2
MFCD00023584
benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
CHEBI:31268
C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2

Specifications

189°C
25g
C13H11NO2
benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2
213.236
CHEBI:31268
Crystal-Lump at 20°C
Benzyl Nicotinate
Undesignated
94-44-0
MFCD00023584
KVYGGMBOZFWZBQ-UHFFFAOYSA-N
benzyl pyridine-3-carboxylate
7191
213.24
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0957

RTECSNumber : QT0850000

TSCA : Yes