Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl n-butyl ether, 97%

$209.45 - $209.45

Chemical Identifiers

Chemical Name or Material Benzyl n-butyl ether
CAS 588-67-0
Molecular Formula C6H5CH2O(CH2)3CH3
Formula Weight 164.25
MDL Number MFCD00039954
Beilstein 1935234
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1174714

  SDS
alfa aesar™
L1174714
25g Each for $209.45
Description & Specifications

Specifications

Formula Weight 164.25
Chemical Name or Material Benzyl n-butyl ether
Molecular Formula C6H5CH2O(CH2)3CH3
MDL Number MFCD00039954
Assay 97%
Boiling Point 111° to 112°C (23mmHg)
Quantity 25g
CAS 588-67-0
Beilstein 1935234