missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl n-butyl ether, 97%

Manufacturer:  Alfa Aesar™ L1174714

 View more versions of this product

Catalog No. AAL1174714



Specifications

Specifications

Benzyl n-butyl ether
25g
C11H16O
1935234
MAYUYFCAPVDYBQ-UHFFFAOYSA-N
butoxymethylbenzene
61134
97%
111°C to 112°C (23mmHg)
588-67-0
MFCD00039954
benzyl butyl ether, ether, benzyl butyl, butyl benzyl ether, benzyl n-butyl ether, benzene, butoxymethyl, butoxymethyl benzene, n-butyl benzyl ether, unii-8483x8zo8n, ether, benzyl n-butyl, butoxyphenylmethane
CCCCOCC1=CC=CC=C1
164.248
164.25

Chemical Identifiers

588-67-0
164.248
MAYUYFCAPVDYBQ-UHFFFAOYSA-N
61134
CCCCOCC1=CC=CC=C1
C11H16O
MFCD00039954
benzyl butyl ether, ether, benzyl butyl, butyl benzyl ether, benzyl n-butyl ether, benzene, butoxymethyl, butoxymethyl benzene, n-butyl benzyl ether, unii-8483x8zo8n, ether, benzyl n-butyl, butoxyphenylmethane
butoxymethylbenzene
Documents