Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl methyl sulfone, 98+%

$377.12 - $377.12

Chemical Identifiers

Chemical Name or Material Benzyl methyl sulfone
CAS 3112-90-1
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00025074
Beilstein 1864579
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0423318

  SDS
alfa aesar™
L0423318
50g Each for $377.12
Description & Specifications

Specifications

Formula Weight 170.23
Chemical Name or Material Benzyl methyl sulfone
Assay ≥98%
Quantity 50g
Molecular Formula C8H10O2S
Odor Odorless
CAS 3112-90-1
Beilstein 1864579
Melting Point 124° to 127°C
MDL Number MFCD00025074