missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl Methyl Malonate, TCI America™

Manufacturer:  TCI America M097425G

 View more versions of this product

Catalog No. M097425G



Specifications

Specifications

Benzyl Methyl Malonate
25g
C11H12O4
benzyl methyl malonate, 1-benzyl 3-methyl propanedioate, malonic acid 1-benzyl 3-methyl ester, propanedioic acid, methyl phenylmethyl ester, benzylmethyl malonate, methyl benzyl malonate, acmc-1b0zd, benzyl methyl propanedioate, ksc493e1f, benzyl methyl malonate
COC(=O)CC(=O)OCC1=CC=CC=C1
208.213
208.21
≥96.0% (GC)
Yellow
52267-39-7
MFCD00008461
IAUZDBFOEWAQFE-UHFFFAOYSA-N
3-O-benzyl 1-O-methyl propanedioate
572135
Clear Liquid at 20°C

Chemical Identifiers

52267-39-7
208.213
IAUZDBFOEWAQFE-UHFFFAOYSA-N
572135
COC(=O)CC(=O)OCC1=CC=CC=C1
C11H12O4
MFCD00008461
benzyl methyl malonate, 1-benzyl 3-methyl propanedioate, malonic acid 1-benzyl 3-methyl ester, propanedioic acid, methyl phenylmethyl ester, benzylmethyl malonate, methyl benzyl malonate, acmc-1b0zd, benzyl methyl propanedioate, ksc493e1f, benzyl methyl malonate
3-O-benzyl 1-O-methyl propanedioate
Safety and Handling

Safety and Handling

TSCA : No

Documents