missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl Methyl Ether 99%, ACROS Organics™

$385.95 - $385.95

Chemical Identifiers

CAS 538-86-3
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula CH3OCH2C6H5
Formula Weight 122.17
MDL Number MFCD00025901
Synonym (Methoxymethyl)benzene
Chemical Name or Material Benzyl methyl ether, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC402142500 SDS Thermo Fisher Scientific
402142500
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC402141000 SDS Thermo Fisher Scientific
402141000
100g Each for $385.95
Specifications Description & Specifications

Chemical Identifiers

538-86-3
100.0
C8H10O
122.17
(Methoxymethyl)benzene
≥98.5%
98.5
99%
CH3OCH2C6H5
MFCD00025901
Benzyl methyl ether, 99%

Specifications

122.17
250g
15, 1143
(Methoxymethyl)benzene
C8H10O
538-86-3
CH3OCH2C6H5
Authentic
Benzyl methyl ether, 99%
Liquid
Clear colorless to slightly yellow
100.0
MFCD00025901
98.5
99%
1.5010 to 1.5030 (20°C, 589nm)
Glass bottle
≥98.5%

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-705-7

TSCA : TSCA