Please login to your online account to display your discounted pricing

Benzyl Methyl Ether 99%, ACROS Organics™


Quantity 250g

Manufacturer: thermo fisher scientific  402142500

 View more versions of this product

Catalog No. AC402142500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 122.17
Physical Form Liquid
Quantity 250g
Color Clear colorless to slightly yellow
Merck Index 15, 1143
TSCA TSCA
EINECS Number 208-705-7
CAS Max % 100.0
Synonym (Methoxymethyl)benzene
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Chemical Name or Material Benzyl methyl ether
MDL Number MFCD00025901
Name Note 99%
Molecular Formula C8H10O
CAS Min % 98.5
CAS 538-86-3
Assay Percent Range 99%
Linear Formula CH3OCH2C6H5
Refractive Index 1.5010 to 1.5030 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Benzyl methyl ether
Name Note 99%
CAS 538-86-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula CH3OCH2C6H5
Formula Weight 122.17
MDL Number MFCD00025901
Synonym (Methoxymethyl)benzene
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents