Please login to your online account to display your discounted pricing

Benzyl Methyl Ether 99%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  402141000

 View more versions of this product

Catalog No. AC402141000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 122.17
Quantity 100g
Color Clear colorless to slightly yellow
Merck Index 15, 1143
EINECS Number 208-705-7
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material Benzyl methyl ether
Refractive Index 1.5010 to 1.5030 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C8H10O
CAS 538-86-3
Synonym (Methoxymethyl)benzene
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Linear Formula CH3OCH2C6H5
MDL Number MFCD00025901
CAS Min % 98.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Packaging Glass bottle

Specifications

Chemical Name or Material Benzyl methyl ether
Name Note 99%
CAS 538-86-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula CH3OCH2C6H5
Formula Weight 122.17
MDL Number MFCD00025901
Synonym (Methoxymethyl)benzene
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents