missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl mercaptan, polymer supported, 1% cross-linked, 100-200 mesh, 2.0-4.0 mmol/g on poly(styrene-divinylbenzene)

Manufacturer:  Alfa Aesar™ 4446618

 View more versions of this product

Catalog No. AA4446618



Specifications

Specifications

Benzyl mercaptan
Pink to White
Polymer supported, 1% cross-linked, 2.0 to 4.0 mmol/g on poly(styrene-divinylbenzene)
C7H8S
alpha-toluenethiol
C1=CC=C(C=C1)CS
124.201
Beads
100 to 200 Mesh
50g
100-53-8
MFCD01863867
UENWRTRMUIOCKN-UHFFFAOYSA-N
phenylmethanethiol
7509

Chemical Identifiers

100-53-8
124.201
UENWRTRMUIOCKN-UHFFFAOYSA-N
7509
C1=CC=C(C=C1)CS
C7H8S
MFCD01863867
alpha-toluenethiol
phenylmethanethiol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents