missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl mercaptan, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2008136

 View more versions of this product

Catalog No. AAB2008136



Description

Description

A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Benzyl mercaptan
-30°C
194°C to 195°C
0.99
C7H8S
MFCD00004867,MFCD00801588,MFCD01863867
UN2810
Air Sensitive
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
UENWRTRMUIOCKN-UHFFFAOYSA-N
phenylmethanethiol
7509
99%
100-53-8
1.053
70°C (158°F)
Stench
1.576
500 g
605864
14,9322
Not miscible or difficult to mix in water.
SCC1=CC=CC=C1
124.20
124.21
Liquid

Chemical Identifiers

100-53-8
124.20
UENWRTRMUIOCKN-UHFFFAOYSA-N
7509
SCC1=CC=CC=C1
C7H8S
MFCD00004867,MFCD00801588,MFCD01863867
benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
phenylmethanethiol
Safety and Handling

Safety and Handling

GHS H Statement
H330-H227-H302
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.

P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P304+P340-P305+P351+P338-P310-P330-P370+P378q-P501c

H227-H302-H319-H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 202-862-5

RTECSNumber : XT8650000

TSCA : Yes

storageNote1 : Air Sensitive

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only