missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl L-(+)-Mandelate, TCI America™

$105.42 - $307.09

Chemical Identifiers

CAS 62173-99-3
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00274325
InChI Key JFKWZVQEMSKSBU-AWEZNQCLSA-N
Synonym L-(+)-Mandelic Acid Benzyl Ester
PubChem CID 667423
IUPAC Name benzyl (2S)-2-hydroxy-2-phenylacetate
SMILES C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13555G View Documents TCI America
M13555G
5g Each for $105.42
 
M135525G View Documents TCI America
M135525G
25g Each for $307.09
 
Specifications

Chemical Identifiers

62173-99-3
242.274
JFKWZVQEMSKSBU-AWEZNQCLSA-N
667423
C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
C15H14O3
MFCD00274325
L-(+)-Mandelic Acid Benzyl Ester
benzyl (2S)-2-hydroxy-2-phenylacetate

Specifications

White
5g
C15H14O3
L-(+)-Mandelic Acid Benzyl Ester
C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O
242.274
242.27
≥98.0% (GC)
107°C
62173-99-3
MFCD00274325
JFKWZVQEMSKSBU-AWEZNQCLSA-N
benzyl (2S)-2-hydroxy-2-phenylacetate
667423
Crystal-Powder at 20°C
Benzyl L-(+)-Mandelate
Safety and Handling

Safety and Handling

TSCA : No