missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl L-lactate, 98%

Manufacturer:  Alfa Aesar™ L1464514

 View more versions of this product

Catalog No. AAL1464514



Description

Description

Applications
Benzyl L-lactate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with oxidizing agents.
Specifications

Specifications

Benzyl L-lactate
−12.9° (c=3.1 in Methanol)
25g
C10H12O3
2048925
ZYTLPUIDJRKAAM-QMMMGPOBSA-N
benzyl (2S)-2-hydroxypropanoate
11052238
98%
>110°C (230°F)
105°C to 109°C (0.1mmHg)
56777-24-3
MFCD00209546
2s-lactic acid benzyl ester
CC(C(=O)OCC1=CC=CC=C1)O
180.203
180.2

Chemical Identifiers

56777-24-3
180.203
ZYTLPUIDJRKAAM-QMMMGPOBSA-N
11052238
CC(C(=O)OCC1=CC=CC=C1)O
C10H12O3
MFCD00209546
2s-lactic acid benzyl ester
benzyl (2S)-2-hydroxypropanoate
Documents