Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl isovalerate, 98%

$209.34 - $209.34

Chemical Identifiers

Chemical Name or Material Benzyl isovalerate
CAS 103-38-8
Molecular Formula (CH3)2CHCH2CO2CH2(C6H5)
Formula Weight 192.26
MDL Number MFCD00048348
Synonym Isovaleric acid benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2485230

 
alfa aesar™
B2485230
250g Each for $209.34
Description & Specifications

Specifications

Formula Weight 192.26
Chemical Name or Material Benzyl isovalerate
TSCA TSCA
Assay 98%
Boiling Point 245°C
Quantity 250g
Molecular Formula (CH3)2CHCH2CO2CH2(C6H5)
MDL Number MFCD00048348
CAS 103-38-8
Synonym Isovaleric acid benzyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.