missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl Isopropenyl Ether [Hydroxyl-Protecting Agent], TCI America™

$247.50 - $656.95

Chemical Identifiers

CAS 32783-20-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00059913
InChI Key ZIFHFFOAEFKJJL-UHFFFAOYSA-N
Synonym 2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether
PubChem CID 560989
IUPAC Name prop-1-en-2-yloxymethylbenzene
SMILES CC(=C)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12571ML View Documents TCI America
B12571ML
1mL Each for $247.50
 
B12575ML View Documents TCI America
B12575ML
5mL Each for $656.95
 
Specifications

Chemical Identifiers

32783-20-3
148.205
ZIFHFFOAEFKJJL-UHFFFAOYSA-N
560989
CC(=C)OCC1=CC=CC=C1
C10H12O
MFCD00059913
2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether
prop-1-en-2-yloxymethylbenzene

Specifications

192°C
1mL
C10H12O
2-Benzyloxy-1-propene, Isopropenyl Benzyl Ether
CC(=C)OCC1=CC=CC=C1
148.205
148.21
≥97.0% (GC)
Colorless
32783-20-3
MFCD00059913
ZIFHFFOAEFKJJL-UHFFFAOYSA-N
prop-1-en-2-yloxymethylbenzene
560989
Clear Liquid at 20°C
Benzyl Isopropenyl Ether [Hydroxyl-Protecting Agent]
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator