missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl isocyanate, 99+%, ACROS Organics™

$110.02 - $110.02

Chemical Identifiers

CAS 3173-56-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00009701
InChI Key YDNLNVZZTACNJX-UHFFFAOYSA-N
Synonym 1-isocyanatomethyl benzene, benzene, isocyanatomethyl, benzyl isocyanate, benzylisocyanate, isocyanatomethyl benzene, isocyanic acid benzyl ester, phenylmethanisocyanate, phenylmethyl isocyanate, unii-8i2li357gq
PubChem CID 76639
IUPAC Name isocyanatomethylbenzene
SMILES O=C=NCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC210760050
SDS View Documents
Thermo Fisher Scientific
210760050
5g Glass bottle
Each for $110.02
 
AC210760250
SDS View Documents
Thermo Fisher Scientific
210760250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

3173-56-6
133.15
YDNLNVZZTACNJX-UHFFFAOYSA-N
76639
O=C=NCC1=CC=CC=C1
C8H7NO
MFCD00009701
1-isocyanatomethyl benzene, benzene, isocyanatomethyl, benzyl isocyanate, benzylisocyanate, isocyanatomethyl benzene, isocyanic acid benzyl ester, phenylmethanisocyanate, phenylmethyl isocyanate, unii-8i2li357gq
isocyanatomethylbenzene

Specifications

1.0700g/mL
Authentic
1.5241 to 1.5261
101°C to 104°C (33.0mmHg)
5g
99.0
99+%
C6H5CH2NCO
12, 1059
1-isocyanatomethyl benzene, benzene, isocyanatomethyl, benzyl isocyanate, benzylisocyanate, isocyanatomethyl benzene, isocyanic acid benzyl ester, phenylmethanisocyanate, phenylmethyl isocyanate, unii-8i2li357gq
O=C=NCC1=CC=CC=C1
133.15
133.15
99+%
45°C
Glass bottle
1.07
Colorless to Yellow
3173-56-6
100.0
C8H7NO
MFCD00009701
13,30; 01,1005
YDNLNVZZTACNJX-UHFFFAOYSA-N
isocyanatomethylbenzene
76639
Liquid
Benzyl isocyanate, 99+%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if swallowed.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-640-9

SDS
Product Certifications