missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl isocyanate, 99+%, ACROS Organics™

Manufacturer:  Acros Organics 210760050

 View more versions of this product

Catalog No. AC210760050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Benzyl isocyanate
45°C
Glass bottle
1.07
Colorless to Yellow
99+%
99.0
99+%
C6H5CH2NCO
12, 1059
1-isocyanatomethyl benzene
C1=CC=C(C=C1)CN=C=O
133.15
133.15
≥99.0%
1.0700g/mL
Authentic
1.5241 to 1.5261
101°C to 104°C (33.0mmHg)
5g
3173-56-6
100.0
C8H7NO
MFCD00009701
13,30; 01,1005
YDNLNVZZTACNJX-UHFFFAOYSA-N
isocyanatomethylbenzene
76639
Liquid

Chemical Identifiers

3173-56-6
133.15
YDNLNVZZTACNJX-UHFFFAOYSA-N
76639
C1=CC=C(C=C1)CN=C=O
C8H7NO
MFCD00009701
1-isocyanatomethyl benzene
isocyanatomethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if swallowed.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-640-9

SDS Documents