Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl isobutyrate, 98+%

$62.39 - $62.39

Chemical Identifiers

Chemical Name or Material Benzyl isobutyrate
CAS 103-28-6
Molecular Formula (CH3)2CHCO2CH2C6H5
Formula Weight 178.23
MDL Number MFCD00048315
Beilstein 1869299
Synonym Isobutyric acid benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0593018

  SDS
alfa aesar™
L0593018
50g Each for $62.39
Description & Specifications

Specifications

Formula Weight 178.23
Chemical Name or Material Benzyl isobutyrate
Molecular Formula (CH3)2CHCO2CH2C6H5
CAS 103-28-6
MDL Number MFCD00048315
TSCA TSCA
Assay ≥98%
Boiling Point 228° to 230°C
Quantity 50g
Synonym Isobutyric acid benzyl ester
Beilstein 1869299
Flash Point >100°C (212°F)
RTECS Number NQ4550000