Please login to your online account to display your discounted pricing

Alfa Aesar™ Benzyl formate, 97%

$30.14 - $78.20

Chemical Identifiers

Chemical Name or Material Benzyl formate
CAS 104-57-4
Molecular Formula HCO2CH2C6H5
Formula Weight 136.14
MDL Number MFCD00014128
Beilstein 2041319
Merck Index 14,1134
Synonym Formic acid benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAL1340514

  SDS
alfa aesar™
L1340514
Each for $30.14

AAL1340522

  SDS
alfa aesar™
L1340522
Each for $78.20
Description & Specifications

Specifications

Formula Weight 136.14
CAS 104-57-4
Assay 97%
Molecular Formula HCO2CH2C6H5
Chemical Name or Material Benzyl formate
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00014128
Beilstein 2041319
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Quantity 25g
Boiling Point 201° to 203°C
Synonym Formic acid benzyl ester
TSCA TSCA
RTECS Number LQ5400000
Health Hazard 1 Warning
Merck Index 14,1134
Flash Point 94°C (201°F)

Applications
Benzyl formate is a used as a flavoring agent (Spicy, jasmine, fruity, floral, ) and used as a perfumery fixer. It is used as a solvent for cellulose. Benzyl formate, is used in synthetic applications of flash vacuum pyrolysis.

Solubility
Insoluble in water, soluble in organic solvents, oils.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.